Learn More
Hampton Research NATRIX 2 NO40 185ML
SDP
Catalog No. 501695
Encompass
HR2-917-40/Natrix 2 #40 (H04), 185 ml - 0.08 M Potassium chloride, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 7.0, 50% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-169-5 Supplier Hampton Research Supplier No. HR291740
HR2-917-40/Natrix 2 #40 (H04), 185 ml - 0.08 M Potassium chloride, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 7.0, 50% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.