Learn More
Hampton Research NATRIX 2 NO41 185ML
SDP
Catalog No. 501696
Encompass
HR2-917-41/Natrix 2 #41 (H05), 185 ml - 0.08 M Potassium chloride, 0.04 M Sodium cacodylate trihydrate pH 7.0, 60% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-169-6 Supplier Hampton Research Supplier No. HR291741
HR2-917-41/Natrix 2 #41 (H05), 185 ml - 0.08 M Potassium chloride, 0.04 M Sodium cacodylate trihydrate pH 7.0, 60% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.