Learn More
Hampton Research NATRIX 2 NO43 185ML
SDP
Catalog No. 501698
Encompass
HR2-917-43/Natrix 2 #43 (H07), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M PIPES pH 7.5, 4% w/v Polyethylene glycol 8,000, 0.001 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-169-8 Supplier Hampton Research Supplier No. HR291743
HR2-917-43/Natrix 2 #43 (H07), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M PIPES pH 7.5, 4% w/v Polyethylene glycol 8,000, 0.001 M Spermine. Please note items from this supplier are non-returnable.