Learn More
Hampton Research NATRIX 2 NO44 185ML
SDP
Catalog No. 501699
Encompass
HR2-917-44/Natrix 2 #44 (H08), 185 ml - 0.015 M Magnesium chloride hexahydrate, 0.002 M Barium chloride dihydrate, 0.05 M PIPES pH 7.5, 7% v/v 2-Propanol, 0.0005 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-169-9 Supplier Hampton Research Supplier No. HR291744
HR2-917-44/Natrix 2 #44 (H08), 185 ml - 0.015 M Magnesium chloride hexahydrate, 0.002 M Barium chloride dihydrate, 0.05 M PIPES pH 7.5, 7% v/v 2-Propanol, 0.0005 M Spermine. Please note items from this supplier are non-returnable.