Learn More
Hampton Research NATRIX 2 NO45 185ML
SDP
Catalog No. 501700
Encompass
HR2-917-45/Natrix 2 #45 (H09), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M PIPES pH 7.5, 10% w/v 1,6-Hexanediol, 0.001 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-170-0 Supplier Hampton Research Supplier No. HR291745
HR2-917-45/Natrix 2 #45 (H09), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M PIPES pH 7.5, 10% w/v 1,6-Hexanediol, 0.001 M Spermine. Please note items from this supplier are non-returnable.