Learn More
Hampton Research NATRIX 2 NO47 185ML
SDP
Catalog No. 501702
Encompass
HR2-917-47/Natrix 2 #47 (H11), 185 ml - 0.2 M Calcium chloride dihydrate, 0.05 M HEPES sodium pH 7.5, 28% v/v Polyethylene glycol 400, 0.002 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-170-2 Supplier Hampton Research Supplier No. HR291747
HR2-917-47/Natrix 2 #47 (H11), 185 ml - 0.2 M Calcium chloride dihydrate, 0.05 M HEPES sodium pH 7.5, 28% v/v Polyethylene glycol 400, 0.002 M Spermine. Please note items from this supplier are non-returnable.