Learn More
Hampton Research NATRIX NO01 185ML
SDP
Catalog No. 501704
Encompass
HR2-918-01/Natrix #01 (A1), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M MES monohydrate pH 5.6, 1.8 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-170-4 Supplier Hampton Research Supplier No. HR291801
HR2-918-01/Natrix #01 (A1), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M MES monohydrate pH 5.6, 1.8 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.