Learn More
Hampton Research NATRIX NO02 185ML
SDP
Catalog No. 501705
Encompass
HR2-918-02/Natrix #02 (A2), 185 ml - 0.01 M Magnesium acetate tetrahydrate, 0.05 M MES monohydrate pH 5.6, 2.5 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-170-5 Supplier Hampton Research Supplier No. HR291802
HR2-918-02/Natrix #02 (A2), 185 ml - 0.01 M Magnesium acetate tetrahydrate, 0.05 M MES monohydrate pH 5.6, 2.5 M Ammonium sulfate. Please note items from this supplier are non-returnable.