Learn More
Hampton Research NATRIX NO03 185ML
SDP
Catalog No. 501706
Encompass
HR2-918-03/Natrix #03 (A3), 185 ml - 0.1 M Magnesium acetate tetrahydrate, 0.05 M MES monohydrate pH 5.6, 20 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-170-6 Supplier Hampton Research Supplier No. HR291803
HR2-918-03/Natrix #03 (A3), 185 ml - 0.1 M Magnesium acetate tetrahydrate, 0.05 M MES monohydrate pH 5.6, 20 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.