Learn More
Hampton Research NATRIX NO04 185ML
SDP
Catalog No. 501707
Encompass
HR2-918-04/Natrix #04 (A4), 185 ml - 0.2 M Potassium chloride, 0.01 M Magnesium sulfate heptahydrate, 0.05 M MES monohydrate pH 5.6, 10 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-170-7 Supplier Hampton Research Supplier No. HR291804
HR2-918-04/Natrix #04 (A4), 185 ml - 0.2 M Potassium chloride, 0.01 M Magnesium sulfate heptahydrate, 0.05 M MES monohydrate pH 5.6, 10 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.