Learn More
Hampton Research NATRIX NO05 185ML
SDP
Catalog No. 501708
Encompass
HR2-918-05/Natrix #05 (A5), 185 ml - 0.2 M Potassium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M MES monohydrate pH 5.6, 5 % w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-170-8 Supplier Hampton Research Supplier No. HR291805
HR2-918-05/Natrix #05 (A5), 185 ml - 0.2 M Potassium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M MES monohydrate pH 5.6, 5 % w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.