Learn More
Hampton Research NATRIX NO07 185ML
SDP
Catalog No. 501710
Encompass
HR2-918-07/Natrix #07 (A7), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M MES monohydrate pH 6.0, 15 % v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-171-0 Supplier Hampton Research Supplier No. HR291807
HR2-918-07/Natrix #07 (A7), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M MES monohydrate pH 6.0, 15 % v/v 2-Propanol. Please note items from this supplier are non-returnable.