Learn More
Hampton Research NATRIX NO08 185ML
SDP
Catalog No. 501711
Encompass
HR2-918-08/Natrix #08 (A8), 185 ml - 0.1 M Ammonium acetate, 0.005 M Magnesium sulfate heptahydrate, 0.05 M MES monohydrate pH 6.0, 0.6 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-171-1 Supplier Hampton Research Supplier No. HR291808
HR2-918-08/Natrix #08 (A8), 185 ml - 0.1 M Ammonium acetate, 0.005 M Magnesium sulfate heptahydrate, 0.05 M MES monohydrate pH 6.0, 0.6 M Sodium chloride. Please note items from this supplier are non-returnable.