Learn More
Hampton Research NATRIX NO09 185ML
SDP
Catalog No. 501712
Encompass
HR2-918-09/Natrix #09 (A9), 185 ml - 0.1 M Potassium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M MES monohydrate pH 6.0, 10 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-171-2 Supplier Hampton Research Supplier No. HR291809
HR2-918-09/Natrix #09 (A9), 185 ml - 0.1 M Potassium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M MES monohydrate pH 6.0, 10 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.