Learn More
Hampton Research NATRIX NO10 185ML
SDP
Catalog No. 501713
Encompass
HR2-918-10/Natrix #10 (A10), 185 ml - 0.005 M Magnesium sulfate heptahydrate, 0.05 M MES monohydrate pH 6.0, 5 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-171-3 Supplier Hampton Research Supplier No. HR291810
HR2-918-10/Natrix #10 (A10), 185 ml - 0.005 M Magnesium sulfate heptahydrate, 0.05 M MES monohydrate pH 6.0, 5 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.