Learn More
Hampton Research NATRIX NO11 185ML
SDP
Catalog No. 501714
Encompass
HR2-918-11/Natrix #11 (A11), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 1.0 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-171-4 Supplier Hampton Research Supplier No. HR291811
HR2-918-11/Natrix #11 (A11), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 1.0 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.