Learn More
Hampton Research NATRIX NO12 185ML
SDP
Catalog No. 501715
Encompass
HR2-918-12/Natrix #12 (A12), 185 ml - 0.01 M Magnesium sulfate heptahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 1.8 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-171-5 Supplier Hampton Research Supplier No. HR291812
HR2-918-12/Natrix #12 (A12), 185 ml - 0.01 M Magnesium sulfate heptahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 1.8 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.