Learn More
Hampton Research NATRIX NO13 185ML
SDP
Catalog No. 501716
Encompass
HR2-918-13/Natrix #13 (B1), 185 ml - 0.015 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 1.7 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-171-6 Supplier Hampton Research Supplier No. HR291813
HR2-918-13/Natrix #13 (B1), 185 ml - 0.015 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 1.7 M Ammonium sulfate. Please note items from this supplier are non-returnable.