Learn More
Hampton Research NATRIX NO14 185ML
SDP
Catalog No. 501717
Encompass
HR2-918-14/Natrix #14 (B2), 185 ml - 0.1 M Potassium chloride, 0.025 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 15 % v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-171-7 Supplier Hampton Research Supplier No. HR291814
HR2-918-14/Natrix #14 (B2), 185 ml - 0.1 M Potassium chloride, 0.025 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 15 % v/v 2-Propanol. Please note items from this supplier are non-returnable.