Learn More
Hampton Research NATRIX NO15 185ML
SDP
Catalog No. 501718
Encompass
HR2-918-15/Natrix #15 (B3), 185 ml - 0.04 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 5 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-171-8 Supplier Hampton Research Supplier No. HR291815
HR2-918-15/Natrix #15 (B3), 185 ml - 0.04 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 5 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.