Learn More
Hampton Research NATRIX NO16 185ML
SDP
Catalog No. 501719
Encompass
HR2-918-16/Natrix #16 (B4), 185 ml - 0.04 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 30 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-171-9 Supplier Hampton Research Supplier No. HR291816
HR2-918-16/Natrix #16 (B4), 185 ml - 0.04 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 30 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.