Learn More
Hampton Research NATRIX NO17 185ML
SDP
Catalog No. 501720
Encompass
HR2-918-17/Natrix #17 (B5), 185 ml - 0.2 M Potassium chloride, 0.01 M Calcium chloride dihydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 10 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-172-0 Supplier Hampton Research Supplier No. HR291817
HR2-918-17/Natrix #17 (B5), 185 ml - 0.2 M Potassium chloride, 0.01 M Calcium chloride dihydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 10 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.