Learn More
Hampton Research NATRIX NO18 185ML
SDP
Catalog No. 501721
Encompass
HR2-918-18/Natrix #18 (B6), 185 ml - 0.01 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 1.3 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-172-1 Supplier Hampton Research Supplier No. HR291818
HR2-918-18/Natrix #18 (B6), 185 ml - 0.01 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 1.3 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.