Learn More
Hampton Research NATRIX NO19 185ML
SDP
Catalog No. 501722
Encompass
HR2-918-19/Natrix #19 (B7), 185 ml - 0.01 M Magnesium sulfate heptahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-172-2 Supplier Hampton Research Supplier No. HR291819
HR2-918-19/Natrix #19 (B7), 185 ml - 0.01 M Magnesium sulfate heptahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.