Learn More
Hampton Research NATRIX NO20 185ML
SDP
Catalog No. 501723
Encompass
HR2-918-20/Natrix #20 (B8), 185 ml - 0.1 M Ammonium acetate, 0.015 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 10 % v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-172-3 Supplier Hampton Research Supplier No. HR291820
HR2-918-20/Natrix #20 (B8), 185 ml - 0.1 M Ammonium acetate, 0.015 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 10 % v/v 2-Propanol. Please note items from this supplier are non-returnable.