Learn More
Hampton Research NATRIX NO22 185ML
SDP
Catalog No. 501725
Encompass
HR2-918-22/Natrix #22 (B10), 185 ml - 0.08 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 15 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-172-5 Supplier Hampton Research Supplier No. HR291822
HR2-918-22/Natrix #22 (B10), 185 ml - 0.08 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 15 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.