Learn More
Hampton Research NATRIX NO24 185ML
SDP
Catalog No. 501727
Encompass
HR2-918-24/Natrix #24 (B12), 185 ml - 0.2 M Ammonium acetate, 0.01 M Calcium chloride dihydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 10 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-172-7 Supplier Hampton Research Supplier No. HR291824
HR2-918-24/Natrix #24 (B12), 185 ml - 0.2 M Ammonium acetate, 0.01 M Calcium chloride dihydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 10 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.