Learn More
Hampton Research NATRIX NO25 185ML
SDP
Catalog No. 501728
Encompass
HR2-918-25/Natrix #25 (C1), 185 ml - 0.08 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 30 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-172-8 Supplier Hampton Research Supplier No. HR291825
HR2-918-25/Natrix #25 (C1), 185 ml - 0.08 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 30 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.