Learn More
Hampton Research NATRIX NO26 185ML
SDP
Catalog No. 501729
Encompass
HR2-918-26/Natrix #26 (C2), 185 ml - 0.2 M Potassium chloride, 0.1 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 10 % w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-172-9 Supplier Hampton Research Supplier No. HR291826
Add to Cart
Add to Cart
HR2-918-26/Natrix #26 (C2), 185 ml - 0.2 M Potassium chloride, 0.1 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 10 % w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.