Learn More
Hampton Research NATRIX NO27 185ML
SDP
Catalog No. 501730
Encompass
HR2-918-27/Natrix #27 (C3), 185 ml - 0.2 M Ammonium acetate, 0.01 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 30 % w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-173-0 Supplier Hampton Research Supplier No. HR291827
HR2-918-27/Natrix #27 (C3), 185 ml - 0.2 M Ammonium acetate, 0.01 M Magnesium acetate tetrahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 30 % w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.