Learn More
Hampton Research NATRIX NO28 185ML
SDP
Catalog No. 501731
Encompass
HR2-918-28/Natrix #28 (C4), 185 ml - 0.05 M Magnesium sulfate hydrate, 0.05 M HEPES Sodium pH 7.0, 1.6 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-173-1 Supplier Hampton Research Supplier No. HR291828
HR2-918-28/Natrix #28 (C4), 185 ml - 0.05 M Magnesium sulfate hydrate, 0.05 M HEPES Sodium pH 7.0, 1.6 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.