Learn More
Hampton Research NATRIX NO29 185ML
SDP
Catalog No. 501732
Encompass
HR2-918-29/Natrix #29 (C5), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 4.0 M Lithium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-173-2 Supplier Hampton Research Supplier No. HR291829
HR2-918-29/Natrix #29 (C5), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 4.0 M Lithium chloride. Please note items from this supplier are non-returnable.