Learn More
Hampton Research NATRIX NO30 185ML
SDP
Catalog No. 501733
Encompass
HR2-918-30/Natrix #30 (C6), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-173-3 Supplier Hampton Research Supplier No. HR291830
HR2-918-30/Natrix #30 (C6), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.