Learn More
Hampton Research NATRIX NO31 185ML
SDP
Catalog No. 501734
Encompass
HR2-918-31/Natrix #31 (C7), 185 ml - 0.005 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 25 % v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.
Catalog No. 50-173-4 Supplier Hampton Research Supplier No. HR291831
HR2-918-31/Natrix #31 (C7), 185 ml - 0.005 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 25 % v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.