Learn More
Hampton Research NATRIX NO32 185ML
SDP
Supplier:  Hampton Research HR291832
Encompass
HR2-918-32/Natrix #32 (C8), 185 ml - 0.2 M Potassium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 1.7 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-173-5