Learn More
Hampton Research NATRIX NO33 185ML
SDP
Catalog No. 501736
Encompass
HR2-918-33/Natrix #33 (C9), 185 ml - 0.2 M Ammonium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 2.5 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-173-6 Supplier Hampton Research Supplier No. HR291833
HR2-918-33/Natrix #33 (C9), 185 ml - 0.2 M Ammonium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 2.5 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.