Learn More
Hampton Research NATRIX NO34 185ML
SDP
Catalog No. 501737
Encompass
HR2-918-34/Natrix #34 (C10), 185 ml - 0.1 M Potassium chloride, 0.005 M Magnesium sulfate hydrate, 0.05 M HEPES Sodium pH 7.0, 15 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-173-7 Supplier Hampton Research Supplier No. HR291834
HR2-918-34/Natrix #34 (C10), 185 ml - 0.1 M Potassium chloride, 0.005 M Magnesium sulfate hydrate, 0.05 M HEPES Sodium pH 7.0, 15 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.