Learn More
Hampton Research NATRIX NO35 185ML
SDP
Catalog No. 501738
Encompass
HR2-918-35/Natrix #35 (C11), 185 ml - 0.1 M Potassium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 5 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-173-8 Supplier Hampton Research Supplier No. HR291835
HR2-918-35/Natrix #35 (C11), 185 ml - 0.1 M Potassium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 5 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.