Learn More
Hampton Research NATRIX NO36 185ML
SDP
Catalog No. 501739
Encompass
HR2-918-36/Natrix #36 (C12), 185 ml - 0.1 M Potassium chloride, 0.01 M Calcium chloride dihydrate, 0.05 M HEPES Sodium pH 7.0, 10 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-173-9 Supplier Hampton Research Supplier No. HR291836
HR2-918-36/Natrix #36 (C12), 185 ml - 0.1 M Potassium chloride, 0.01 M Calcium chloride dihydrate, 0.05 M HEPES Sodium pH 7.0, 10 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.