Learn More
Hampton Research NATRIX NO37 185ML
SDP
Catalog No. 501740
Encompass
HR2-918-37/Natrix #37 (D1), 185 ml - 0.2 M Potassium chloride, 0.025 M Magnesium sulfate hydrate, 0.05 M HEPES Sodium pH 7.0, 20 % v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.
Catalog No. 50-174-0 Supplier Hampton Research Supplier No. HR291837
HR2-918-37/Natrix #37 (D1), 185 ml - 0.2 M Potassium chloride, 0.025 M Magnesium sulfate hydrate, 0.05 M HEPES Sodium pH 7.0, 20 % v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.