Learn More
Hampton Research NATRIX NO38 185ML
SDP
Catalog No. 501741
Encompass
HR2-918-38/Natrix #38 (D2), 185 ml - 0.2 M Ammonium acetate, 0.15 M Magnesium acetate tetrahydrate, 0.05 M HEPES Sodium pH 7.0, 5 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-174-1 Supplier Hampton Research Supplier No. HR291838
HR2-918-38/Natrix #38 (D2), 185 ml - 0.2 M Ammonium acetate, 0.15 M Magnesium acetate tetrahydrate, 0.05 M HEPES Sodium pH 7.0, 5 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.