Learn More
Hampton Research NATRIX NO39 185ML
SDP
Catalog No. 501742
Encompass
HR2-918-39/Natrix #39 (D3), 185 ml - 0.1 M Ammonium acetate, 0.02 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 5 % w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-174-2 Supplier Hampton Research Supplier No. HR291839
HR2-918-39/Natrix #39 (D3), 185 ml - 0.1 M Ammonium acetate, 0.02 M Magnesium chloride hexahydrate, 0.05 M HEPES Sodium pH 7.0, 5 % w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.