Learn More
Hampton Research NATRIX NO40 185ML
SDP
Catalog No. 501743
Encompass
HR2-918-40/Natrix #40 (D4), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-174-3 Supplier Hampton Research Supplier No. HR291840
HR2-918-40/Natrix #40 (D4), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 1.6 M Ammonium sulfate. Please note items from this supplier are non-returnable.