Learn More
Hampton Research NATRIX NO41 185ML
SDP
Catalog No. 501744
Encompass
HR2-918-41/Natrix #41 (D5), 185 ml - 0.1 M Potassium chloride, 0.015 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 10 % v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.
Catalog No. 50-174-4 Supplier Hampton Research Supplier No. HR291841
HR2-918-41/Natrix #41 (D5), 185 ml - 0.1 M Potassium chloride, 0.015 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 10 % v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.