Learn More
Hampton Research NATRIX NO42 185ML
SDP
Catalog No. 501745
Encompass
HR2-918-42/Natrix #42 (D6), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 5 % v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-174-5 Supplier Hampton Research Supplier No. HR291842
HR2-918-42/Natrix #42 (D6), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 5 % v/v 2-Propanol. Please note items from this supplier are non-returnable.