Learn More
Hampton Research NATRIX NO43 185ML
SDP
Catalog No. 501746
Encompass
HR2-918-43/Natrix #43 (D7), 185 ml - 0.05 M Ammonium acetate, 0.01 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 10 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-174-6 Supplier Hampton Research Supplier No. HR291843
HR2-918-43/Natrix #43 (D7), 185 ml - 0.05 M Ammonium acetate, 0.01 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 10 % v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.