Learn More
Hampton Research NATRIX NO44 185ML
SDP
Catalog No. 501747
Encompass
HR2-918-44/Natrix #44 (D8), 185 ml - 0.2 M Potassium chloride, 0.05 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 10 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-174-7 Supplier Hampton Research Supplier No. HR291844
HR2-918-44/Natrix #44 (D8), 185 ml - 0.2 M Potassium chloride, 0.05 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 7.5, 10 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.