Learn More
Hampton Research NATRIX NO45 185ML
SDP
Catalog No. 501748
Encompass
HR2-918-45/Natrix #45 (D9), 185 ml - 0.025 M Magnesium sulfate hydrate, 0.05 M TRIS hydrochloride pH 8.5, 1.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-174-8 Supplier Hampton Research Supplier No. HR291845
HR2-918-45/Natrix #45 (D9), 185 ml - 0.025 M Magnesium sulfate hydrate, 0.05 M TRIS hydrochloride pH 8.5, 1.8 M Ammonium sulfate. Please note items from this supplier are non-returnable.