Learn More
Hampton Research NATRIX NO46 185ML
SDP
Catalog No. 501749
Encompass
HR2-918-46/Natrix #46 (D10), 185 ml - 0.005 M Magnesium sulfate hydrate, 0.05 M TRIS hydrochloride pH 8.5, 2.9 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-174-9 Supplier Hampton Research Supplier No. HR291846
HR2-918-46/Natrix #46 (D10), 185 ml - 0.005 M Magnesium sulfate hydrate, 0.05 M TRIS hydrochloride pH 8.5, 2.9 M 1,6-Hexanediol. Please note items from this supplier are non-returnable.