Learn More
Hampton Research NATRIX NO47 185ML
SDP
Catalog No. 501750
Encompass
HR2-918-47/Natrix #47 (D11), 185 ml - 0.1 M Potassium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 8.5, 30 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-175-0 Supplier Hampton Research Supplier No. HR291847
HR2-918-47/Natrix #47 (D11), 185 ml - 0.1 M Potassium chloride, 0.01 M Magnesium chloride hexahydrate, 0.05 M TRIS hydrochloride pH 8.5, 30 % v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.